Mercury(II) nitrate monohydrate, 98+%, Thermo Scientific Chemicals
Mercury(II) nitrate monohydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury(II) nitrate monohydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 213140500
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.50 
Save 13.85 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.50 
Save 13.85 (15%)
Add to cart
Mercury(II) nitrate monohydrate, 98+%, Thermo Scientific Chemicals
Catalog number213140500
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.50 
Save 13.85 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namemercury(2+) hydrate dinitrate
Molecular FormulaH2HgN2O7
InChI KeyKVICROHOONHSRH-UHFFFAOYSA-N
SMILESO.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol)342.62
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to slightly yellow
Appearance (Form)Adhering crystalline powder or crystals
Titration with NH4SCN98.0 to 102.0 %
Chloride (Cl)=<20 ppm
Sulfate (SO4)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only