Thermo Scientific Chemicals

Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific Chemicals

Catalog number: 213181000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific Chemicals

Catalog number: 213181000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 213181000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54678-23-8
IUPAC Name
(methylsulfanyl)methane; bromocopper
Molecular Formula
C2H6BrCuS
InChI Key
PMHQVHHXPFUNSP-UHFFFAOYSA-M
SMILES
[Cu]Br.CSC
Appearance (Color)
Off-white to beige to green to grey
Titration Iodometric
>=98.5 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text