1-Hexene, 97%
1-Hexene, 97%
1-Hexene, 97%
Thermo Scientific Chemicals

1-Hexene, 97%

1-Hexene, 97%, C6H12, CAS Number-592-41-6, dialene 6, butyl ethylene, 1-hexene, butylethylene, 1-n-hexene, hexylene, hexene-1, hexene, n-hexene, unii-b38zz8c206, 1L, 96% min. (GC), 01, 215, 62.0 deg.C to 65.0 deg.C, 100.0, 96.0, CHEBI:24579, Colorless, 0.6780g/mL, 209-753-1, -26 deg.C, 84.15 | CAS: 592-41-6 | C6H12 | 84.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number 213211000
Price (USD)
31.65
Special offer
Online exclusive
Ends: 26-Sep-2025
36.50
Save 4.85 (13%)
Each
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 26-Sep-2025
36.50
Save 4.85 (13%)
Each
Add to cart
Chemical Identifiers
CAS592-41-6
IUPAC Namehex-1-ene
Molecular FormulaC6H12
InChI KeyLIKMAJRDDDTEIG-UHFFFAOYSA-N
SMILESCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3867 to 1.3887 (20°C, 589 nm)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link