Sodium ethoxide, pure, 21% in ethanol, Thermo Scientific Chemicals
Sodium ethoxide, pure, 21% in ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium ethoxide, pure, 21% in ethanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
250 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 213220010
Price (USD)/ Each
128.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
151.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
128.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
151.00 
Save 22.35 (15%)
Add to cart
Sodium ethoxide, pure, 21% in ethanol, Thermo Scientific Chemicals
Catalog number213220010
Price (USD)/ Each
128.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
151.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium ethanolate
Molecular FormulaC2H5NaO
InChI KeyQDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES[Na+].CC[O-]
Molecular Weight (g/mol)68.05
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
AppearanceMay darken on storage
NaOH + Na2CO3 (as NaOH)=<0.5 %
Appearance (Color)Yellow to brown
Titration with HCl20.0 to 22.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use