Thermo Scientific Chemicals

Sodium ethoxide, pure, 21% in ethanol, Thermo Scientific Chemicals

Catalog number: 213221000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium ethoxide, pure, 21% in ethanol, Thermo Scientific Chemicals

Catalog number: 213221000
100 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 213221000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-52-6
IUPAC Name
sodium ethanolate
Molecular Formula
C2H5NaO
InChI Key
QDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES
[Na+].CC[O-]
Appearance (Form)
Solution
Appearance (Color)
Yellow to brown
Titration with HCl
20.0 to 22.0 %
Appearance
May darken on storage
NaOH + Na2CO3 (as NaOH)
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text