Thermo Scientific Chemicals

Methyl 4-cyanobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 213280250
25 g, Each
Thermo Scientific Chemicals

Methyl 4-cyanobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 213280250
25 g, Each
Quantity
Catalog number: 213280250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1129-35-7
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
66°C to 70°C
GC
>=97.5 %
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text