n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
Thermo Scientific Chemicals

n-Butyllithium, 2.5M solution in hexanes, AcroSeal™

CAS: 109-72-8 | C4H9Li | 64.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
50 mL
100 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 213351000
Price (USD)
63.65
Special offer
Online exclusive
Ends: 26-Sep-2025
78.90
Save 15.25 (19%)
Each
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 26-Sep-2025
78.90
Save 15.25 (19%)
Each
Add to cart
Chemical Identifiers
CAS109-72-8
IUPAC Namebutyllithium
Molecular FormulaC4H9Li
InChI KeyMZRVEZGGRBJDDB-UHFFFAOYSA-N
SMILES[Li]CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy colorless to yellow
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Titration Active base22.0 to 24.0 %
Titration Active base2.37 to 2.59 M
View more
Ask our AI about this Product

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link