Thermo Scientific Chemicals

n-Butyllithium, 2.5M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 213358000
800 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

n-Butyllithium, 2.5M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 213358000
800 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 213358000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-72-8
IUPAC Name
butyllithium
Molecular Formula
C4H9Li
InChI Key
MZRVEZGGRBJDDB-UHFFFAOYSA-N
SMILES
[Li]CCCC
Appearance (Color)
Clear to hazy colorless to yellow
Appearance (Form)
Solution
Appearance
May develop some turbidity or precipitate
Titration Active base
22.0 to 24.0 %
Titration Active base
2.37 to 2.59 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text