Thermo Scientific Chemicals

Chromium(III) chloride hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 213365000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Chromium(III) chloride hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 213365000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 213365000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10060-12-5
IUPAC Name
chromium(3+) hexahydrate trichloride
Molecular Formula
Cl3CrH12O6
InChI Key
LJAOOBNHPFKCDR-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
Appearance (Color)
Dark green
Appearance (Form)
Crystalline powder
Titration with Na2S2O3
>=97.5 %
Iron (Fe)
=<300 ppm
Sulfate (SO4)
=<500 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text