Thermo Scientific™

EZ-Link™ Hydrazide-Biotin

Catalog number: 21339
Have Questions?
Thermo Scientific™

EZ-Link™ Hydrazide-Biotin

Catalog number: 21339
Have Questions?
Catalog Number
21339
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
21339100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific EZ-Link Hydrazide-Biotin is the shortest and simplest hydrazide-activated biotinylation reagent for labeling glycoproteins and other carbohydrate-containing compounds having oxidizable sugars or aldehydes.

Features of EZ-Link Hydrazide-Biotin:

Glycoprotein labeling—biotinylate glycosylated proteins at sialic acid residues for detection or purification using streptavidin probes or resins
Cell surface labeling—biotinylate and isolate cell surface glycoproteins
Aldehyde-reactive—reacts with aldehydes formed by periodate-oxidation of sugar groups
Hydrazide-activated—perform reactions at pH 4 to 6 in buffers such as sodium acetate
Irreversible—forms semi-permanent hydrazone bonds; spacer arm cannot be cleaved
Solubility—usually dissolved in DMSO before further dilution in aqueous buffers
Spacer arm length—15.7Å

This biotin hydrazide reagent enables simple and efficient biotin labeling of polyclonal antibodies and other glycoproteins. Mild oxidation of antibodies with sodium periodate produces reactive aldehydes on the carbohydrate moieties of the Fc portion that can be modified by hydrazides. This approach is advantageous for labeling antibodies because biotinylation occurs only at the sites of glycosylation, which are primarily in the Fc region of the antibody, far from the antigen binding site.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.

Biotinylation reagents differ in reactivity, length, solubility, cell permeability and cleavability. Hydrazides and alkoxyamines are two types of carbonyl-reactive groups. Hydrazides (—NH-NH2) react specifically with aldehyde groups in slightly acidic conditions to form hydrazone linkages; these can be further reduced to stable secondary amine bonds using sodium cyanoborohydride (Part No. 44892). The reaction is more efficient in the presence of aniline (Part No. 88944). Alternatively, hydrazides can be conjugated to carboxylic acids using EDC carbodiimide chemistry.

Reactive aldehyde groups can be generated in glycoproteins and other polysaccharide compounds by oxidation of constituent sugar diols using sodium periodiate (Part No. 20504). Sialic acid residues are common components of protein glycosylation and are easily converted to aldehydes with 1 mM NaIO4.

Related Products
EZ-Link™ Biotin-LC-Hydrazide
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Hydrazide-Biotin
Quantity
100 mg
Product Line
EZ-Link™
Cell Permeability
Cell-Permeant
Chemical Reactivity
Carboxylic Acid, Ketone, Aldehyde
Reactive Moiety
Amine, Hydrazide
Solubility
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye
Biotin
Spacer
Short
Label Type
Biotin & Analogs

Contents & Storage

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text