Thermo Scientific Chemicals

Antimony(III) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 213471000
100 g, Each
Thermo Scientific Chemicals

Antimony(III) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 213471000
100 g, Each
Quantity
Catalog number: 213471000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-64-4
IUPAC Name
diantimony(3+) trioxidandiide
Molecular Formula
O3Sb2
InChI Key
GHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Sb+3].[Sb+3]
Appearance (Form)
Powder
Arsenic (As)
=<500 ppm
Lead (Pb)
=<400 ppm
Particle size
=<0.01 % Residue on 45 µm sieve
Average particle size
0.8 to 1.0 µm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text