Manganese(IV) oxide, 80-85%, powder, Thermo Scientific Chemicals
Manganese(IV) oxide, 80-85%, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(IV) oxide, 80-85%, powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 213490010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namedioxomanganese
Molecular FormulaMnO2
InChI KeyNUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILESO=[Mn]=O
Molecular Weight (g/mol)86.94
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with Na2S2O3>=80 %
Loss on drying=<0.5 % (105°C)
Insoluble matter=<10 % (in HCl)
Iron (Fe)=<1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only