Informational:Looking for a higher binding capacity magnetic bead that provides easy conjugation of proteins for your custom affinity purification experiments? Try our Pierce NHS-Activated Magnetic Beads. Pierce Magnetic beads accelerate throughput through validated and optimized automated methods and provide premium performance with manual methods. Please contact us with any questions.
Thermo Scientific™

MagnaBind™ Amine Derivatized Beads

Catalog number: 21352
Have Questions?
Thermo Scientific™

MagnaBind™ Amine Derivatized Beads

Catalog number: 21352
Have Questions?
Catalog Number
21352
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
213525 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific™ MagnaBind™ Beads provide a convenient method for magnetic separation of antibodies, antigens, lectins, enzymes, nucleic acids and cells using affinity binding. To remove the MagnaBind Beads from the suspension, an external magnetic field is used.

MagnaBind Amine Derivatized Beads are supplied as an aqueous suspension of magnetic iron oxide beads coated with amine groups for covalent coupling of molecules using a variety of crosslinkers.

Features of MagnaBind Amine Derivatized Beads:

Composition: Silanized iron oxide
Magnetization: 25-35EMU/g
Type of Magnetization: Superparamagnetic (no magnetic memory)
Surface Area: >100m2/g
Bead Size: 1-4μm diameter
Settling Rate: 4% in 30 minutes
Effective Density: 2.5g/mL
Number of Beads: 1 × 108 beads/mg
pH Stability: Aqueous solution, above pH 4.0
Concentration: ∼50 mg/mL

Applications:
• Cell sorting using either positive or negative selection
• Protein purification or immunoassays using either direct or indirect methods
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
MagnaBind Amine Derivatized Beads
Color
Brown
For Use With (Application)
Affinity Purification
Product Type
Amine Derivatized Bead
Product Line
MagnaBind™
Surface Functionality
Amine
Diameter (Metric)
1 to 4 μm

Contents & Storage

Upon receipt store product at 4°C. Do not freeze product. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text