4-(Trifluoromethyl)benzyl alcohol, 99%, Thermo Scientific Chemicals
4-(Trifluoromethyl)benzyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzyl alcohol, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog number 213570050
Price (USD)/ Each
55.20
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
55.20
Add to cart
4-(Trifluoromethyl)benzyl alcohol, 99%, Thermo Scientific Chemicals
Catalog number213570050
Price (USD)/ Each
55.20
-
Add to cart
Chemical Identifiers
CAS349-95-1
IUPAC Name[4-(trifluoromethyl)phenyl]methanol
Molecular FormulaC8H7F3O
InChI KeyMOOUWXDQAUXZRG-UHFFFAOYSA-N
SMILESOCC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4590 to 1.4610 (20°C, 589 nm)
Appearance (Color)Clear colorless to light brown
Appearance (Form)Liquid after melting
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only