Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water
Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water
Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water
Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water
Thermo Scientific Chemicals

Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water

CAS: 21351-79-1 | CsHO | 149.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 213601000
Price (USD)
102.65
Special offer
Online exclusive
121.00
Save 18.35 (15%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
121.00
Save 18.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS21351-79-1
IUPAC Namecaesium(1+) hydroxide
Molecular FormulaCsHO
InChI KeyHUCVOHYBFXVBRW-UHFFFAOYSA-M
SMILES[OH-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace analysisTrace analysis values are on dry salt basis
Trace analysisTrace analysis values are on dry salt basis
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467092Certificate of AnalysisJan 31, 2025213601000, 213605000
A0454686Certificate of AnalysisAug 25, 2023213601000, 213605000
A0452253Certificate of AnalysisMay 08, 2023213601000, 213605000
A0451522Certificate of AnalysisMay 02, 2023213601000, 213605000
A0397705Certificate of AnalysisDec 15, 2022213601000, 213605000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection