Thermo Scientific Chemicals

Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water, Thermo Scientific Chemicals

Catalog number: 213605000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Cesium hydroxide, 99.9%, (trace metal basis), for analysis, 50 wt% solution in water, Thermo Scientific Chemicals

Catalog number: 213605000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 213605000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21351-79-1
IUPAC Name
caesium(1+) hydroxide
Molecular Formula
CsHO
InChI Key
HUCVOHYBFXVBRW-UHFFFAOYSA-M
SMILES
[OH-].[Cs+]
Additional info
Trace analysis values are on dry salt basis
Sodium (Na)
=<100 ppm
Potassium (K)
=<200 ppm
Rubidium (Rb)
=<10 ppm
Magnesium (Mg)
=<1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text