[Bis(trifluoroacetoxy)iodo]benzene, 98%, Thermo Scientific Chemicals
[Bis(trifluoroacetoxy)iodo]benzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

[Bis(trifluoroacetoxy)iodo]benzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 213670100
Price (USD)/ Each
72.65
Online exclusive
81.10 
Save 8.45 (10%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
81.10 
Save 8.45 (10%)
Add to cart
[Bis(trifluoroacetoxy)iodo]benzene, 98%, Thermo Scientific Chemicals
Catalog number213670100
Price (USD)/ Each
72.65
Online exclusive
81.10 
Save 8.45 (10%)
-
Add to cart
Chemical Identifiers
CAS2712-78-9
IUPAC Namephenyl[(2,2,2-trifluoroacetyl)oxy]-λ³-iodanyl 2,2,2-trifluoroacetate
Molecular FormulaC10H5F6IO4
InChI KeyPEZNEXFPRSOYPL-UHFFFAOYSA-N
SMILESFC(F)(F)C(=O)O[I](OC(=O)C(F)(F)F)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point121°C to 125°C
Appearance (Color)White to light yellow
Titration Iodometric>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagent
RUO – Research Use Only