Thermo Scientific Chemicals

[Bis(trifluoroacetoxy)iodo]benzene, 98%, Thermo Scientific Chemicals

Catalog number: 213670500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

[Bis(trifluoroacetoxy)iodo]benzene, 98%, Thermo Scientific Chemicals

Catalog number: 213670500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 213670500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2712-78-9
IUPAC Name
phenyl[(2,2,2-trifluoroacetyl)oxy]-λ³-iodanyl 2,2,2-trifluoroacetate
Molecular Formula
C10H5F6IO4
InChI Key
PEZNEXFPRSOYPL-UHFFFAOYSA-N
SMILES
FC(F)(F)C(=O)O[I](OC(=O)C(F)(F)F)C1=CC=CC=C1
Infrared spectrum
Conforms
Melting point
121°C to 125°C
Appearance (Color)
White to light yellow
Titration Iodometric
>=97.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text