Methyl 1-cyclohexene-1-carboxylate, 98%, Thermo Scientific Chemicals
Methyl 1-cyclohexene-1-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 1-cyclohexene-1-carboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 213710250
Price (USD)/ Each
436.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
436.00
Add to cart
Methyl 1-cyclohexene-1-carboxylate, 98%, Thermo Scientific Chemicals
Catalog number213710250
Price (USD)/ Each
436.00
-
Add to cart
Chemical Identifiers
CAS18448-47-0
IUPAC Namemethyl cyclohex-1-ene-1-carboxylate
Molecular FormulaC8H12O2
InChI KeyKXPWRCPEMHIZGU-UHFFFAOYSA-N
SMILESCOC(=O)C1=CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4750 to 1.4790 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only