Thermo Scientific Chemicals

Pyridinium p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 213741000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Pyridinium p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 213741000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 213741000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24057-28-1
IUPAC Name
4-methylbenzene-1-sulfonic acid; pyridine
Molecular Formula
C12H13NO3S
InChI Key
ZDYVRSLAEXCVBX-UHFFFAOYSA-N
SMILES
C1=CC=NC=C1.CC1=CC=C(C=C1)S(O)(=O)=O
Appearance (Color)
White to light beige
Melting point
112°C to 119°C
Appearance (Form)
Crystalline powder or crystals
Titration with NaOH
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text