Glycyrrhizic acid monoammonium salt, 98+%, Thermo Scientific Chemicals
Glycyrrhizic acid monoammonium salt, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glycyrrhizic acid monoammonium salt, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number 213751000
Price (USD)/ Each
193.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
228.00 
Save 34.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
193.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
228.00 
Save 34.35 (15%)
Add to cart
Glycyrrhizic acid monoammonium salt, 98+%, Thermo Scientific Chemicals
Catalog number213751000
Price (USD)/ Each
193.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
228.00 
Save 34.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2S,3S,4S,5R,6S)-6-{[(3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl]oxy}-5-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3,4-dihydroxyoxane-2-carboxylic acid amine
Molecular FormulaC42H65NO16
InChI KeyILRKKHJEINIICQ-OOFFSTKBSA-N
SMILESN.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O
Molecular Weight (g/mol)839.97
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Heavy metals=<20 ppm
Specific optical rotationon anhydrous substance
Appearance (Color)White to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only