Thermo Scientific Chemicals

Cobalt(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals

Catalog number: 214130050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

Cobalt(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals

Catalog number: 214130050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 214130050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7646-79-9
IUPAC Name
λ²-cobalt(2+) dichloride
Molecular Formula
Cl2Co
InChI Key
GVPFVAHMJGGAJG-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Co++]
Titration Complexometric
>=96.0 %
Lead (Pb)
=<100 ppm
Appearance (Form)
Powder
Nickel (Ni)
=<2000 ppm
Iron (Fe)
=<500 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text