Thermo Scientific Chemicals

Tungsten(VI) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 214211000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tungsten(VI) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 214211000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 214211000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-35-8
IUPAC Name
trioxotungsten
Molecular Formula
O3W
InChI Key
ZNOKGRXACCSDPY-UHFFFAOYSA-N
SMILES
O=[W](=O)=O
Appearance (Form)
Powder
Appearance (Color)
Yellow to yellow-green
Assay
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text