Thermo Scientific Chemicals

Methyl 3-amino-2-thiophenecarboxylate, 99%, Thermo Scientific Chemicals

Catalog number: 214300500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-amino-2-thiophenecarboxylate, 99%, Thermo Scientific Chemicals

Catalog number: 214300500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 214300500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
22288-78-4
IUPAC Name
methyl 3-aminothiophene-2-carboxylate
Molecular Formula
C6H7NO2S
InChI Key
TWEQNZZOOFKOER-UHFFFAOYSA-N
SMILES
COC(=O)C1=C(N)C=CS1
Appearance (Form)
Crystalline powder
Titration with HClO4
>=98.5 %
Melting point
64°C to 66°C
Infrared spectrum
Conforms
Appearance (Color)
Beige to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text