Thermo Scientific™

EZ-Link™ Sulfo NHS-SS Biotinylation Kit

Catalog number: 21445
Have Questions?
Thermo Scientific™

EZ-Link™ Sulfo NHS-SS Biotinylation Kit

Catalog number: 21445
Have Questions?
Catalog Number
21445
Unit Size
10 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2144510 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific EZ-Link Sulfo NHS-SS Biotinylation Kit contains sufficient reagents for 10 biotinylation reactions (e.g., 1–10 mg antibody per reaction). The EZ-Link Sulfo-NHS-SS-Biotin included in the kit is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.

Features of EZ-Link Sulfo-NHS-SS-Biotin:

Protein labeling – biotinylate antibodies to facilitate immobilization, purification or detection
Cell surface labeling – biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
Cleavable—disulfide bond in spacer arm allows the biotin label to be removed using reducing agents such as DTT; only a small sulfhydryl group remains attached to the molecule
Soluble – charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
Medium length—spacer arm (total length added to target) is 24.3 angstroms; it consists of the native biotin valeric acid group extended by a 7-atom chain

Sulfo-NHS-SS-Biotin is a thiol-cleavable amine-reactive biotinylation reagent that contains an extended spacer arm to reduce steric hindrances associated with avidin binding. This reagent is water-soluble, enabling biotinylation to be performed in the absence of organic solvents such as DMSO or DMF for applications that cannot tolerate solvents or are complicated by their inclusion. The compound is particularly useful for labeling and purifying cell surface proteins, because (a) its sulfonate group prevents it from permeating cell membranes and (b) its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency, and performance for the intended research applications.

N-Hydroxysulfosuccinimide (NHS) esters of biotin are the most popular type of biotinylation reagent. NHS-activated biotins react efficiently with primary amino groups (-NH2) in alkaline buffers to form stable amide bonds. Proteins (e.g., antibodies) typically have several primary amines that are available as targets for labeling, including the side chain of lysine (K) residues and the N-terminus of each polypeptide.

Varieties of biotin NHS-ester reagents differ in length, solubility, cell permeability and cleavability. Non-sulfonated NHS-biotins are cell permeable but must be dissolved in organic solvent such as DMSO or DMF. Sulfo-NHS biotins (and those with pegylated spacers) are directly water soluble but not membrane permeable. Varieties containing disulfide bonds can be cleaved using reducing agents, enabling the biotin group to be disconnected from the labeled protein.

Related Products
EZ-Link™ Sulfo-NHS-SS-Biotin
EZ-Link™ Micro Sulfo-NHS-SS-Biotinylation Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
EZ-Link Sulfo-NHS-SS-Biotinylation Kit
Product Type
Sulfo-NHS-SS Biotinylation Kit
Labeling Scale
1 to 10 mg
Product Line
EZ-Link™
Solubility
Water
Label or Dye
Biotin
Labeling Target
Antibodies, Proteins

Contents & Storage

Sufficient For: 10 labeling reactions, each with 1 to 10 mg of antibody
• EZ-Link Sulfo-NHS-SS-Biotin, 25 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 5 mL, 10 columns
• HABA Solution, 1 mL
• Avidin, 10 mg

Store avidin at -20°C. Store biotin reagent desiccated at 4°C. Store remaining kit components at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text