4'-(Trifluoromethyl)acetophenone, 99%, Thermo Scientific Chemicals
4'-(Trifluoromethyl)acetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-(Trifluoromethyl)acetophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 214450250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS709-63-7
IUPAC Name1-[4-(trifluoromethyl)phenyl]ethan-1-one
Molecular FormulaC9H7F3O
InChI KeyHHAISVSEJFEWBZ-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
Appearance (Color)White to light yellow
Infrared spectrumConforms
Melting point30°C to 33°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only