Thermo Scientific Chemicals

2,6-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 214460050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

2,6-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 214460050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 214460050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24821-22-5
IUPAC Name
2,6-bis(trifluoromethyl)benzoic acid
Molecular Formula
C9H4F6O2
InChI Key
XZNLSDPNMNWCRE-UHFFFAOYSA-N
SMILES
OC(=O)C1=C(C=CC=C1C(F)(F)F)C(F)(F)F
HPLC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Melting point
136°C to 140°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text