Thermo Scientific™

EZ-Link™ NHS-PEG4 Biotinylation Kit

Catalog number: 21455
Have Questions?
Thermo Scientific™

EZ-Link™ NHS-PEG4 Biotinylation Kit

Catalog number: 21455
Have Questions?
Catalog Number
21455
Unit Size
8 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
214558 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific EZ-Link NHS-PEG4 Biotinylation Kit contains EZ-Link NHS-PEG4-Biotin and accessory reagents required for 8 1–10 mg protein labeling reactions. EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.

Features of EZ-Link NHS-PEG4-Biotin:

Protein labeling—biotinylate antibodies or other proteins for detection or purification using streptavidin probes or resins
Amine-reactive—reacts with primary amines (-NH2), such as the side-chain of lysines (K) or the amino-termini of polypeptides
Pegylated – spacer arm contains a hydrophilic, 4-unit, polyethylene glycol (PEG) group
Enhances solubility – pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
Irreversible—forms permanent amide bonds; spacer arm cannot be cleaved
Long reach – spacer arm (total length added to target) is 29 angstroms; this reduces steric hindrance when binding to avidin molecules

NHS-PEG4-Biotin is a long (29.0Å), pegylated, water-soluble, NHS-ester biotinylation reagent to label amines and maximize solubility of antibodies and other proteins. The N-hydroxysuccinimide ester (NHS) group reacts specifically and efficiently with lysine and N-terminal amino groups to form stable amide bonds. The hydrophilic polyethylene glycol (PEG) spacer arm imparts water solubility that is transferred to the biotinylated molecule, thus reducing aggregation of labeled proteins stored in solution. The PEG spacer arm also gives the reagent a long and flexible connection to minimize steric hindrance for binding to avidin molecules.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.

N-Hydroxysulfosuccinimide (NHS) esters of biotin are the most popular type of biotinylation reagent. NHS-activated biotins react efficiently with primary amino groups (-NH2) in alkaline buffers to form stable amide bonds. Proteins (e.g., antibodies) typically have several primary amines that are available as targets for labeling, including the side chain of lysine (K) residues and the N-terminus of each polypeptide.

Varieties of biotin NHS-ester reagents differ in length, solubility, cell permeability and cleavability. Non-sulfonated NHS-biotins are cell permeable but must be dissolved in organic solvent such as DMSO or DMF. Sulfo-NHS biotins (and those with pegylated spacers) are directly water soluble but not membrane permeable. Varieties containing disulfide bonds can be cleaved using reducing agents, enabling the biotin group to be disconnected from the labeled protein.

Related Products
EZ-Link™ NHS-PEG4-Biotin
EZ-Link™ Micro NHS-PEG4-Biotinylation Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
EZ-Link NHS-PEG4 Biotinylation Kit
Product Type
NHS-PEG4 Biotinylation Kit
Quantity
8 Reaction Kit
Labeling Scale
1 to 10 mg
Product Line
EZ-Link™
Solubility
Water
Label or Dye
Biotin
Spacer
Pegylated
Labeling Target
Antibodies, Proteins

Contents & Storage

Sufficient For: 8 labeling reactions, each with 1 to 10 mg of antibody
• EZ-Link NHS-PEG4-Biotin, 8 x 2 mg
• PBS Pack (makes 500mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 5 mL, 10 columns
• HABA Solution, 1 mL
• Avidin, 10mg

Store avidin and microtubes of biotin reagent at -20°C. Store PBS at room temperature. Store remaining kit components at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text