2,6-Diphenylphenol, 97%, Thermo Scientific Chemicals
2,6-Diphenylphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Diphenylphenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 214740050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-phenyl-[1,1'-biphenyl]-2-ol
Molecular FormulaC18H14O
InChI KeyATGFTMUSEPZNJD-UHFFFAOYSA-N
SMILESOC1=C(C=CC=C1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)246.31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point98°C to 103°C
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only