Thermo Scientific Chemicals

4-Amino-2-mercaptopyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: 214770050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Amino-2-mercaptopyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: 214770050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 214770050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
333-49-3
IUPAC Name
6-amino-1,2-dihydropyrimidine-2-thione
Molecular Formula
C4H5N3S
InChI Key
DCPSTSVLRXOYGS-UHFFFAOYSA-N
SMILES
NC1=CC=NC(=S)N1
Appearance (Color)
Yellow to brown
Appearance (Form)
Powder
HPLC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text