Molecular sieves 3A, powder <50 micron, Thermo Scientific Chemicals
Molecular sieves 3A, powder &lt;50 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves 3A, powder <50 micron, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
View all
Plastic Drum
Catalog number 214790025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS308080-99-1
IUPAC NameMolecular sieves 3A
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Particle size=<50 µm
Residual water=<3 % (550°C, 2 h)
Appearance (Color)White
pH=<11.4 (10 % suspension in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only