Thermo Scientific Chemicals

Molecular sieves 3A, powder <50 micron, Thermo Scientific Chemicals

Catalog number: 214795000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Molecular sieves 3A, powder <50 micron, Thermo Scientific Chemicals

Catalog number: 214795000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 214795000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
308080-99-1
IUPAC Name
Molecular sieves 3A
MDL Number
MFCD00131613
Appearance (Form)
Powder
Particle size
=<50 µm
Residual water
=<3 % (550°C, 2 h)
Appearance (Color)
White
pH
=<11.4 (10 % suspension in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text