Thermo Scientific Chemicals

Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals

Catalog number: 214800100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals

Catalog number: 214800100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 214800100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
70955-01-0
IUPAC Name
Molecular sieves
Synonym
2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL Number
MFCD00131613
Water adsorption
>=21 (50% rH, 25°C, 1 bar)
Appearance (Color)
White to beige
Bulk density
300 to 400 g/l
Particle size
=<10 % (>=50 µm)
Loss on ignition
=<1.5 % (550°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text