Molecular sieves 4A, powder <50 micron
Molecular sieves 4A, powder &lt;50 micron
Molecular sieves 4A, powder &lt;50 micron
Molecular sieves 4A, powder &lt;50 micron
Thermo Scientific Chemicals

Molecular sieves 4A, powder <50 micron

CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 214805000
Price (USD)
83.65
Online Exclusive
92.50
Save 8.85 (10%)
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
83.65
Online Exclusive
92.50
Save 8.85 (10%)
Each
Add to cart
Chemical Identifiers
CAS70955-01-0
IUPAC NameMolecular sieves
Synonym2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Water adsorption>=21 (50% rH, 25°C, 1 bar)
Appearance (Color)White to beige
Bulk density300 to 400 g/l
Particle size=<10 % (>=50 µm)
Loss on ignition=<1.5 % (550°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only

Share catalog number, name or link