Molecular sieves 5A, powder <50 micron, Thermo Scientific Chemicals
Molecular sieves 5A, powder &lt;50 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves 5A, powder <50 micron, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 g
Catalog number 214815000
Price (USD)/ Each
66.65
Online exclusive
73.90 
Save 7.25 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
73.90 
Save 7.25 (10%)
Add to cart
Molecular sieves 5A, powder <50 micron, Thermo Scientific Chemicals
Catalog number214815000
Price (USD)/ Each
66.65
Online exclusive
73.90 
Save 7.25 (10%)
-
Add to cart
Chemical Identifiers
CAS69912-79-4
IUPAC NameMolecular sieves 5A
MDL NumberMFCD00147627
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Water adsorption>=23 % (50%rH, 20°C, 24h)
pH10 to 10.8 (10 % suspension in water)
Residual water=<1.5 % (550°C, 2 h)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only