Sodium bisulfate, 92%, Tech., granular, Thermo Scientific Chemicals
Sodium bisulfate, 92%, Tech., granular, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium bisulfate, 92%, Tech., granular, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number 214850025
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
Add to cart
Sodium bisulfate, 92%, Tech., granular, Thermo Scientific Chemicals
Catalog number214850025
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7681-38-1
IUPAC Namesodium hydrogen sulfate
Molecular FormulaHNaO4S
InChI KeyWBHQBSYUUJJSRZ-UHFFFAOYSA-M
SMILES[Na+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration with NaOH>=37.6 % (equivalent H2SO4)
Appearance (Form)Granules
Iron (Fe)=<200 ppm
Titration with NaOH>=90 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only