Sodium bisulfate, 92%, technical
Sodium bisulfate, 92%, technical
Sodium bisulfate, 92%, technical
Sodium bisulfate, 92%, technical
Thermo Scientific Chemicals

Sodium bisulfate, 92%, technical

CAS: 7681-38-1 | HNaO4S | 120.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 214850250
Price (USD)
31.65
Special offer
Online exclusive
36.80
Save 5.15 (14%)
Each
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
36.80
Save 5.15 (14%)
Each
Add to cart
Chemical Identifiers
CAS7681-38-1
IUPAC Namesodium hydrogen sulfate
Molecular FormulaHNaO4S
InChI KeyWBHQBSYUUJJSRZ-UHFFFAOYSA-M
SMILES[Na+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=90 %
Titration with NaOH>=37.6 % (equivalent H2SO4)
Appearance (Color)White to light yellow
Appearance (Form)Granules or crystals or crystalline powder
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link