Thermo Scientific Chemicals

Sodium hexanitrocobaltate(III), Thermo Scientific Chemicals

Catalog number: 214860250
25 g, Each
Thermo Scientific Chemicals

Sodium hexanitrocobaltate(III), Thermo Scientific Chemicals

Catalog number: 214860250
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 214860250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13600-98-1
IUPAC Name
trisodium hexanitrocobalttris(ylium)
Molecular Formula
CoN6Na3O12
InChI Key
BZFWEBJDBATXME-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[Na+].[O-][N+](=O)[Co+3]([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)[N+]([O-])=O
Appearance (Color)
Orange to orange-brown
Appearance (Form)
Powder
Suitability
(for K determination) Passes test
Insoluble matter
=<0.02 % (in dilute acetic acid)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text