Thermo Scientific Chemicals

Nickel(II) bromide hydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 214871000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Nickel(II) bromide hydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 214871000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 214871000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
207569-11-7
IUPAC Name
nickel(2+) dibromide
Molecular Formula
Br2Ni
InChI Key
IPLJNQFXJUCRNH-UHFFFAOYSA-L
SMILES
[Ni++].[Br-].[Br-]
Titration Argentometric
>=97.5 % (on dry substance)
Iron (Fe)
=<20 ppm
Cobalt (Co)
=<100 ppm
Potassium (K)
=<20 ppm
Appearance (Color)
Orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text