Thermo Scientific Chemicals

Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 214980050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 214980050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 214980050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
206986-79-0
IUPAC Name
N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide; bis(acetic acid)
Molecular Formula
C26H38Cl2N10O4
InChI Key
WDRFFJWBUDTUCA-UHFFFAOYSA-N
SMILES
CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1
Titration with HClO4
>=97.5 % (On dry substance)
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Loss on drying
=<3.5 % (105°C)
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text