2149E-HR Full specifications
Case of 4800 ART 10, Filtered, Sterile, Hinged Rack 10 μL Hinged Rack Sterile & PCR Ready - Description ART 10, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 Ultra-micro Length (English) 1.8 in. Length (Metric) 4.57cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Ultra-micro For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 4800
2139-HR Full specifications
Case of 4800 ART 10, Filtered, Sterile, Hinged Rack 10 μL Hinged Rack Sterile & PCR Ready 569.55 Online price 821.00
- Description ART 10, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 Length (English) 1.23 in. Length (Metric) 3.13cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 4800
2139-HRPK Full specifications
Pack of 960 ART 10, Filtered, Sterile, Hinged Rack 10 μL Hinged Rack Sterile & PCR Ready 113.91 Online price 187.80
- Description ART 10, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 Length (English) 1.23 in. Length (Metric) 3.13cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 960
2139-05-HR Full specifications
Case of 4800 ART 10, Filtered, Sterile, Hinged Rack 10 μL Hinged Rack Sterile & PCR Ready 655.90 Online price 937.00
- Description ART 10, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 Length (English) 1.23 in. Length (Metric) 3.13cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Low Retention For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 4800
2139-05-HRPK Full specifications
Pack of 960 ART 10, Filtered, Sterile, Hinged Rack 10 μL Hinged Rack Sterile & PCR Ready - Description ART 10, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 Length (English) 1.23 in. Length (Metric) 3.13cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Low Retention For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 960
2149E-HRPK Full specifications
Pack of 960 ART 10, Filtered, Sterile, Hinged Rack 10 μL Hinged Rack Sterile & PCR Ready - Description ART 10, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 Ultra-micro Length (English) 1.8 in. Length (Metric) 4.57cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Ultra-micro For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 960
2149E-05-HR Full specifications
Case of 4800 ART 20E Ultra-micro, Filtered, Sterile, Hinged Rack 10 μL Hinged Rack Sterile & PCR Ready - Description ART 20E Ultra-micro, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 20E Ultra-micro Length (English) 1.8 in. Length (Metric) 4.57cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Low Retention For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 4800
2149E-05-HRPK Full specifications
Pack of 960 ART 20E Ultra-micro, Filtered, Sterile, Hinged Rack 10 μL Hinged Rack Sterile & PCR Ready - Description ART 20E Ultra-micro, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 20E Ultra-micro Length (English) 1.8 in. Length (Metric) 4.57cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Low Retention For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 960
2149P-HR Full specifications
Case of 4800 ART 20P, Filtered, Sterile, Hinged Rack 20 μL Hinged Rack Sterile & PCR Ready 617.80 Online price 821.00
- Description ART 20P, Filtered, Sterile, Hinged Rack Volume (Metric) 20 μL Filtered Filtered Sterility Sterile Model ART 20P Length (English) 2 in. Length (Metric) 5.07cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 4800
2149P-HRPK Full specifications
Pack of 960 ART 20P, Filtered, Sterile, Hinged Rack 20 μL Hinged Rack Sterile & PCR Ready - Description ART 20P, Filtered, Sterile, Hinged Rack Volume (Metric) 20 μL Filtered Filtered Sterility Sterile Model ART 20P Length (English) 2 in. Length (Metric) 5.07cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 960