HEPES sodium salt, 99%
HEPES sodium salt, 99%
Thermo Scientific Chemicals

HEPES sodium salt, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
1 kg
Catalog number 215001000
Price (USD)/ Each
112.65
Online exclusive
132.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
112.65
Online exclusive
132.00 
Save 19.35 (15%)
Add to cart
HEPES sodium salt, 99%
Catalog number215001000
Price (USD)/ Each
112.65
Online exclusive
132.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular FormulaC8H17N2NaO4S
InChI KeyRDZTWEVXRGYCFV-UHFFFAOYSA-M
SMILES[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
Molecular Weight (g/mol)260.28
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
UV(0.1 M in water)
UVat 260 nm A: =<0.04
Titration with HClO4>=98.5 % (on anhydrous substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only