DL-3-Hydroxybutyric acid, sodium salt, 99%
DL-3-Hydroxybutyric acid, sodium salt, 99%
DL-3-Hydroxybutyric acid, sodium salt, 99%
DL-3-Hydroxybutyric acid, sodium salt, 99%
Thermo Scientific Chemicals

DL-3-Hydroxybutyric acid, sodium salt, 99%

CAS: 150-83-4 | C4H7NaO3 | 126.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
21501025025 gGlass bottle
Catalog number 215010250
Price (USD)
99.65
Special offer
Online exclusive
Ends: 26-Sep-2025
123.00
Save 23.35 (19%)
Each
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 26-Sep-2025
123.00
Save 23.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS150-83-4
IUPAC Namesodium 3-hydroxybutanoate
Molecular FormulaC4H7NaO3
InChI KeyNBPUSGBJDWCHKC-UHFFFAOYNA-M
SMILES[Na+].CC(O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point161°C to 167°C (Dec)
Infrared spectrumConforms
Appearance (Color)White
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link