DL-3-Hydroxybutyric acid, sodium salt, 99%, Thermo Scientific Chemicals
DL-3-Hydroxybutyric acid, sodium salt, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-3-Hydroxybutyric acid, sodium salt, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 215011000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS150-83-4
IUPAC Namesodium 3-hydroxybutanoate
Molecular FormulaC4H7NaO3
InChI KeyNBPUSGBJDWCHKC-UHFFFAOYNA-M
SMILES[Na+].CC(O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point161°C to 167°C (Dec)
Appearance (Color)White
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only