Linoleic acid, 99%
Thermo Scientific Chemicals

Linoleic acid, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 215040050
Price (USD)/ Each
58.65
Online exclusive
65.20 
Save 6.55 (10%)
-
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
58.65
Online exclusive
65.20 
Save 6.55 (10%)
Add to cart
Linoleic acid, 99%
Catalog number215040050
Price (USD)/ Each
58.65
Online exclusive
65.20 
Save 6.55 (10%)
-
Add to cart
Chemical Identifiers
CAS60-33-3
IUPAC Name(9Z,12Z)-octadeca-9,12-dienoic acid
Molecular FormulaC18H32O2
InChI KeyOYHQOLUKZRVURQ-HZJYTTRNSA-N
SMILESCCCCCC=C/CC=C/CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4687 to 1.4707 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only