Thermo Scientific Chemicals

Linoleic acid, 99%

Catalog number: 215040250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Linoleic acid, 99%

Catalog number: 215040250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 215040250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-33-3
IUPAC Name
(9Z,12Z)-octadeca-9,12-dienoic acid
Molecular Formula
C18H32O2
InChI Key
OYHQOLUKZRVURQ-HZJYTTRNSA-N
SMILES
CCCCCC=C/CC=C/CCCCCCCC(O)=O
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4687 to 1.4707 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text