Linoleic acid, 99%
Linoleic acid, 99%
Linoleic acid, 99%
Thermo Scientific Chemicals

Linoleic acid, 99%

CAS: 60-33-3 | C18H32O2 | 280.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 215041000
Price (USD)
359.65
Online Exclusive
399.00
Save 39.35 (10%)
Each
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
359.65
Online Exclusive
399.00
Save 39.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS60-33-3
IUPAC Name(9Z,12Z)-octadeca-9,12-dienoic acid
Molecular FormulaC18H32O2
InChI KeyOYHQOLUKZRVURQ-HZJYTTRNSA-N
SMILESCCCCCC=C/CC=C/CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4687 to 1.4707 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection