Thermo Scientific Chemicals

PIPES, disodium salt, 99%, Thermo Scientific Chemicals

Catalog number: 215092500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

PIPES, disodium salt, 99%, Thermo Scientific Chemicals

Catalog number: 215092500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 215092500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
76836-02-7
IUPAC Name
disodium 2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular Formula
C8H16N2Na2O6S2
InChI Key
GMHSTJRPSVFLMT-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
Appearance (Form)
Powder
pH
9.2 to 10.0 (1 % in water at 25°C)
Appearance (Color)
White
Water
=<5 %
UV
(0.1 M in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text