Spermidine trihydrochloride, 99+%, Thermo Scientific Chemicals
Spermidine trihydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Spermidine trihydrochloride, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 215100250
Price (USD)/ Each
340.65
Online exclusive
378.00 
Save 37.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
340.65
Online exclusive
378.00 
Save 37.35 (10%)
Add to cart
Spermidine trihydrochloride, 99+%, Thermo Scientific Chemicals
Catalog number215100250
Price (USD)/ Each
340.65
Online exclusive
378.00 
Save 37.35 (10%)
-
Add to cart
Chemical Identifiers
CAS334-50-9
IUPAC Nametrihydrogen (4-aminobutyl)(3-aminopropyl)amine trichloride
Molecular FormulaC7H22Cl3N3
InChI KeyLCNBIHVSOPXFMR-UHFFFAOYSA-N
SMILES[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Argentometric>=99.0 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only