Thermo Scientific Chemicals

1,4-Diaminobutane dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 215111000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,4-Diaminobutane dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 215111000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 215111000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
333-93-7
IUPAC Name
dihydrogen butane-1,4-diamine dichloride
Molecular Formula
C4H14Cl2N2
InChI Key
XXWCODXIQWIHQN-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NCCCCN
Titration Argentometric
>=99.0 %
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text